3-Cyano-4-fluorophenylboronic acid pinacol ester ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3-Cyano-4-fluorobenzeneboronic acid pinacol ester, 2-(3-Cyano-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Cyano-4-fluorophenylboronic acid Pinacol ester, 3-Cyano-4-fluorophenylboronic acid, pinacol ester, 2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

F1253-1G F1253-5G
TCF1253-1GEA 53 USD
TCF1253-1G TCF1253-5G
3-Cyano-4-fluorophenylboronic acid pinacol ester ≥98.0% (by GC)
3-Cyano-4-fluorophenylboronic acid pinacol ester
Formula: C₁₃H₁₅BFNO₂
MW: 247.08 g/mol
MDL Number: MFCD06795681
CAS Number: 775351-57-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 73.0 to 77.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR