2-Methoxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Methoxybenzeneboronic acid pinacol ester, 2-Methoxyphenylboronic acid, pinacol ester, 2-(2-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

T3708-25G T3708-5G
TCT3708-25GEA 320 USD
TCT3708-25G TCT3708-5G
2-Methoxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-Methoxyphenylboronic acid pinacol ester
Formula: C₁₃H₁₉BO₃
MW: 234.10 g/mol
MDL Number: MFCD05863916
CAS Number: 190788-60-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 84.0 to 88.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR