4-Nitro-2-(trifluoromethyl)benzonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

N1068-25G N1068-5G
TCN1068-25GEA 250 USD
TCN1068-25G TCN1068-5G
4-Nitro-2-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
4-Nitro-2-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₃N₂O₂
MDL Number: MFCD01075729
CAS Number: 320-47-8
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 49.0 to 53.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR