3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) ≥97.0% (by HPLC) for biochemical research
Supplier: TCI America
Synonyms:
Thiazolyl blue tetrazolium bromide, MTT, 2-(4,5-Dimethylthiazol-2-yl)-3,5-diphenyl-2H-tetrazol-3-ium bromide
M3297-1G
M3297-200MG
TCM3297-1GEA
65
USD
TCM3297-1G
TCM3297-200MG
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) ≥97.0% (by HPLC) for biochemical research
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)
Formula:
C₁₈H₁₆N₅BrS MW: 414.33 g/mol |
MDL Number:
MFCD00011964 CAS Number: 298-93-1 EINECS: 206-069-5 |
Specification Test Results
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Functionality test | to pass test (by Abs method) |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...