6-Methoxytryptamine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

M3162-100MG M3162-500MG
TCM3162-100MGEA 110 USD
TCM3162-100MG TCM3162-500MG
6-Methoxytryptamine ≥98.0% (by HPLC, titration analysis)
6-Methoxytryptamine
Formula: C₁₁H₁₄N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00005663
CAS Number: 3610-36-4
EINECS: 222-778-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 141.0 to 145.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR