2-Fluoro-3-methoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

F1218-1G F1218-5G
TCF1218-1GEA 37 USD
TCF1218-1G TCF1218-5G
2-Fluoro-3-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
2-Fluoro-3-methoxybenzoic acid
Formula: C₈H₇FO₃
MDL Number: MFCD02179621
CAS Number: 137654-20-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 160.0 to 164.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR