2-Hydroxy-3-nitro-5-picoline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-Methyl-3-nitro-2-pyridone, 2-Hydroxy-5-methyl-3-nitropyridine, 6-Hydroxy-5-nitro-3-picoline, 5-Methyl-3-nitro-2-pyridinol, 5-Methyl-3-nitro-2(1H)-pyridone

H1483-25G H1483-5G
TCH1483-25GEA 281 USD
TCH1483-25G TCH1483-5G
2-Hydroxy-3-nitro-5-picoline ≥98.0% (by GC, titration analysis)
2-Hydroxy-3-nitro-5-picoline
Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
MDL Number: MFCD02070021
CAS Number: 7464-14-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR