3-Fluorobenzenesulfonamide ≥98.0% (by GC)

Supplier: TCI America
Danger

F1141-1G F1141-5G
TCF1141-1GEA 39 USD
TCF1141-1G TCF1141-5G
3-Fluorobenzenesulfonamide ≥98.0% (by GC)
3-Fluorobenzenesulphonamide
Formula: C₆H₆FNO₂S
MW: 175.18 g/mol
MDL Number: MFCD00042284
CAS Number: 1524-40-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 128.0 to 132.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR