4-Fluoro-3-(trifluoromethyl)benzenesulfonyl chloride ≥98.0% (by GC)

Supplier: TCI America
Danger

F1127-25G F1127-5G
TCF1127-25GEA 255 USD
TCF1127-25G TCF1127-5G
4-Fluoro-3-(trifluoromethyl)benzenesulfonyl chloride ≥98.0% (by GC)
4-Fluoro-3-(trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₃ClF₄O₂S
MDL Number: MFCD01091001
CAS Number: 1682-10-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR