3-Fluoro-2-methylbenzaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3-Fluoro-o-tolualdehyde, 3-Fluoro-2-tolualdehyde, 3-Fluoro-o-tolylaldehyde

F1197-1G F1197-5G
TCF1197-1GEA 25 USD
TCF1197-1G TCF1197-5G
3-Fluoro-2-methylbenzaldehyde ≥98.0% (by GC)
3-Fluoro-2-methylbenzaldehyde
Formula: C₈H₇FO
Boiling Pt: 60 °C
Flash Pt: 81 °C
MDL Number: MFCD00075256
CAS Number: 147624-13-3

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Acid Value max. 5.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR