Ethyl ethoxyacetate ≥95.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Ethyl 2-ethoxyacetate, Ethoxyacetic acid ethyl ester

E1237-25G E1237-5G
TCE1237-25GEA 138 USD
TCE1237-25G TCE1237-5G
Ethyl ethoxyacetate ≥95.0% (by GC)
Ethyl ethoxyacetate
Formula: C₆H₁₂O₃
Boiling Pt: 158 °C
Flash Pt: 50 °C
MDL Number: MFCD00040492
CAS Number: 817-95-8
EINECS: 212-447-0
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR