1,2-Dibromo-4-nitrobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

D5486-1G D5486-5G
TCD5486-1GEA 21 USD
TCD5486-1G TCD5486-5G
1,2-Dibromo-4-nitrobenzene ≥98.0% (by GC)
1,2-Dibromo-4-nitrobenzene
Formula: C₆H₃Br₂NO₂
Boiling Pt: 296 °C
MDL Number: MFCD00055373
CAS Number: 5411-50-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR