Dextrin, Soluble fiber

Supplier: TCI America

Synonyms: Dextrin

D5658-100G D5658-500G
TCD5658-100GEA 30 USD
TCD5658-100G TCD5658-500G
Dextrin, Soluble fiber
Dextrin
Formula: (C₆H₁₂O₆)n
MDL Number: MFCD00081554
CAS Number: 9004-53-9
EINECS: 232-675-4
Merck Index: 14,02953

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Solubility in hot Water transparency
Drying loss max. 10.000 %
Ignition residue (Sulfate) max. 0.5 %
Reducing suger max. 8.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR