3-Amino-5-methylcaproic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

H1587-1G H1587-5G
TCH1587-1GEA 68 USD
TCH1587-1G TCH1587-5G
3-Amino-5-methylcaproic acid ≥98.0% (by titrimetric analysis)
3-Amino-5-methylcaproic acid
Formula: C₇H₁₅NO₂
MDL Number: MFCD01863281
CAS Number: 3653-34-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -1.4 to +1.4 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR