Trimethylsulfoxonium chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

T3794-25G T3794-5G
TCT3794-25GEA 333 USD
TCT3794-25G TCT3794-5G
Trimethylsulfoxonium chloride ≥98.0% (by titrimetric analysis)
Trimethylsulphoxonium chloride
Formula: C₃H₉ClOS
MDL Number: MFCD00011898
CAS Number: 5034-06-0
EINECS: 225-724-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 240.0 to 242.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR