trans-2-Methyl-2-pentenoic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: (E)-2-Methylpent-2-enoic acid

M2814-25G M2814-5G
TCM2814-25GEA 89 USD
TCM2814-25G TCM2814-5G
trans-2-Methyl-2-pentenoic acid ≥97.0% (by GC, titration analysis)
trans-2-Methyl-2-pentenoic acid
Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 125 °C
Flash Pt: 108 °C
MDL Number: MFCD00002655
CAS Number: 16957-70-3
EINECS: 241-026-4
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Colourless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR