α,α,α-Trifluoro-m-xylene ≥98.0% (by GC)

Supplier: TCI America
Warning

M2944-1G M2944-5G
TCM2944-1GEA 22 USD
TCM2944-1G TCM2944-5G
α,α,α-Trifluoro-m-xylene ≥98.0% (by GC)
α,α,α-Trifluoro-m-xylene
Formula: C₈H₇F₃
Flash Pt: 27 °C
MDL Number: MFCD00190105
CAS Number: 401-79-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR