4-Iodo-2-nitroaniline ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
I1029-1G
I1029-5G
TCI1029-1GEA
37
USD
TCI1029-1G
TCI1029-5G
4-Iodo-2-nitroaniline ≥98.0% (by GC)
4-Iodo-2-nitroaniline
Formula:
C₆H₅IN₂O₂ MW: 264.02 g/mol |
MDL Number:
MFCD06808526 CAS Number: 20691-72-9 |
Specification Test Results
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 124.0 to 128.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...