4-(Trifluoromethyl)anilinium chloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

T3833-1G T3833-5G
TCT3833-1GEA 29 USD
TCT3833-1G TCT3833-5G
4-(Trifluoromethyl)anilinium chloride ≥98.0% (by HPLC, titration analysis)
4-(Trifluoromethyl)anilinium chloride
Formula: C₇H₇ClF₃N
Melting Pt: 202 °C
MDL Number: MFCD00007897
CAS Number: 90774-69-9
EINECS: 292-679-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR