2,5-Diiodo-p-xylene ≥98.0% (by GC)

Supplier: TCI America
Warning

D5589-1G D5589-5G
TCD5589-1GEA 37 USD
TCD5589-1G TCD5589-5G
2,5-Diiodo-p-xylene ≥98.0% (by GC)
2,5-Diiodo-p-xylene
Formula: C₈H₈I₂
MDL Number: MFCD00829126
CAS Number: 1124-08-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 102.0 to 106.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR