Tetrachloropyrocatechol ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

Synonyms: Tetrachlorobenzene-1,2-diol, 3,4,5,6-Tetrachloro-1,2-dihydroxybenzene, Tetrachlorocatechol, 3,4,5,6-Tetrachlorocatechol, 3,4,5,6-Tetrachlorobenzene-1,2-diol

T3627-5G T3627-1G
TCT3627-5GEA 289 USD
TCT3627-5G TCT3627-1G
Tetrachloropyrocatechol ≥98.0% (by HPLC, titration analysis)
Tetrachloropyrocatechol
Formula: C₆H₂Cl₄O₂
MW: 247.89 g/mol
MDL Number: MFCD00002190
CAS Number: 1198-55-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 195.0 to 199.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR