4-Fluoro-2-(trifluoromethyl)benzyl bromide ≥97.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 1-(Bromomethyl)-4-fluoro-2-(trifluoromethyl)benzene

F1149-1G F1149-5G
TCF1149-1GEA 39 USD
TCF1149-1G TCF1149-5G
4-Fluoro-2-(trifluoromethyl)benzyl bromide ≥97.0% (by GC)
4-Fluoro-2-(trifluoromethyl)benzyl bromide
Formula: C₈H₅BrF₄
MW: 257.03 g/mol
Flash Pt: 85 °C
MDL Number: MFCD00061176
CAS Number: 206860-48-2
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR