3-Fluoro-5-(trifluoromethyl)benzyl bromide ≥98.0% (by GC)

Supplier: TCI America
Danger

F1131-1G F1131-5G
TCF1131-1GEA 32 USD
TCF1131-1G TCF1131-5G
3-Fluoro-5-(trifluoromethyl)benzyl bromide ≥98.0% (by GC)
3-Fluoro-5-(trifluoromethyl)benzyl bromide
Formula: C₈H₅BrF₄
MW: 257.03
Boiling Pt: 43
MDL Number: MFCD00061175
CAS Number: 239087-09-3
UN: 3265
ADR: 8 III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR