1-(2-Methylphenyl)piperazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

T3404-25G T3404-5G
TCT3404-25GEA 187 USD
TCT3404-25G TCT3404-5G
1-(2-Methylphenyl)piperazine ≥98.0% (by GC, titration analysis)
1-(2-Methylphenyl)piperazine
Formula: C₁₁H₁₆N₂
Boiling Pt: 161 °C
MDL Number: MFCD00040729
CAS Number: 39512-51-1
EINECS: 254-481-9
UN: 3263
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 44.0 to 48.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR