(Ethyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Ethyltriphenylphosphonium chloride

E1336-100G E1336-25G
TCE1336-100GEA 136 USD
TCE1336-100G TCE1336-25G
(Ethyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
(Ethyl)triphenylphosphonium chloride
Formula: C₂₀H₂₀ClP
MW: 326.81 g/mol
MDL Number: MFCD00044699
CAS Number: 896-33-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 241.0 to 245.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR