1-(4-Methoxyphenyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Danger

Synonyms: 1-(4-Methoxyphenyl)thiourea, (4-Methoxyphenyl)thiourea, N-(4-Methoxyphenyl)thiourea, 4-Methoxyphenylthiourea

M2593-25G M2593-5G
TCM2593-25GEA 565 USD
TCM2593-25G TCM2593-5G
1-(4-Methoxyphenyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)
1-(4-Methoxyphenyl)-2-thiourea
Formula: (C₆H₄(OCH₃))NC(S)NH₂
MDL Number: MFCD00004936
CAS Number: 2293-07-4
EINECS: 218-931-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 206.0 to 210.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR