1-(2-Fluorophenyl)-2-thiourea ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

Synonyms: (2-Fluorophenyl)thiourea, N-(2-Fluorophenyl)thiourea, 2-Fluorophenylthiourea

F1055-1G F1055-5G
TCF1055-1GEA 59 USD
TCF1055-1G TCF1055-5G
1-(2-Fluorophenyl)-2-thiourea ≥98.0% (by HPLC, titration analysis)
1-(2-Fluorophenyl)-2-thiourea
Formula: (C₆H₄F)NC(S)NH₂
MW: 170.21 g/mol
MDL Number: MFCD00041148
CAS Number: 656-32-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 144.0 to 148.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR