Silver(I) hexafluorophosphate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Silver hexafluorophosphate

S0981-1G S0981-5G
TCS0981-1GEA 42 USD
TCS0981-1G TCS0981-5G
Silver(I) hexafluorophosphate ≥98.0% (by titrimetric analysis)
Silver(I) hexafluorophosphate
Formula: AgF₆P
MW: 252.83
MDL Number: MFCD00003415
CAS Number: 26042-63-7
EINECS: 247-428-6
UN: 3260
ADR: 8 II

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (Precipitation Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR