1-(2,5-Xylyl)piperazin ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-(2,5-Dimethylphenyl)piperazine

D5659-25G D5659-5G
TCD5659-25GEA 166 USD
TCD5659-25G TCD5659-5G
1-(2,5-Xylyl)piperazin ≥98.0% (by GC, titration analysis)
1-(2,5-Xylyl)piperazin

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 44.0 to 48.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR