5-Iodo-o-toluidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

I1021-25G I1021-5G
TCI1021-25GEA 345 USD
TCI1021-25G TCI1021-5G
5-Iodo-o-toluidine ≥98.0% (by GC, titration analysis)
5-Iodo-o-toluidine
Formula: C₇H₈IN
MDL Number: MFCD00051531
CAS Number: 83863-33-6
EINECS: 281-094-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 48.0 to 52.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR