Pentamethyldisiloxane ≥95.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 1,1,1,3,3-Pentamethyldisiloxane

P1535-25ML P1535-5ML
TCP1535-25MLEA 136 USD
TCP1535-25ML TCP1535-5ML
Pentamethyldisiloxane ≥95.0% (by GC)
Pentamethyldisiloxane
Formula: C₅H₁₆OSi₂
MW: 148.35 g/mol
Boiling Pt: 86 °C
Flash Pt: −19 °C
MDL Number: MFCD00053658
CAS Number: 1438-82-0
EINECS: 215-873-5
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR