Tetrabutylammonium nitrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

T3651-25G T3651-5G
TCT3651-25GEA 89 USD
TCT3651-25G TCT3651-5G
Tetrabutylammonium nitrate ≥98.0% (by titrimetric analysis)
Tetrabutylammonium nitrate
Formula: C₁₆H₃₆N₂O₃
MDL Number: MFCD00043202
CAS Number: 1941-27-1
EINECS: 217-726-0
UN: 1479
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 117.0 to 121.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR