2,4,6-Trimethylphenyl isothiocyanate ≥98.0% (by GC)

Supplier: TCI America
Danger

T3324-25G T3324-5G
TCT3324-25GEA 323 USD
TCT3324-25G TCT3324-5G
2,4,6-Trimethylphenyl isothiocyanate ≥98.0% (by GC)
2,4,6-Trimethylphenyl isothiocyanate
Formula: C₁₀H₁₁NS
Boiling Pt: 75 °C
MDL Number: MFCD00022055
CAS Number: 6095-82-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 61.0 to 65.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR