(3-Iodophenyl)acetonitrile ≥96.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 3-Iodobenzyl cyanide, 2-(3-Iodophenyl)acetonitrile

I1002-1G I1002-5G
TCI1002-1GEA 107 USD
TCI1002-1G TCI1002-5G
(3-Iodophenyl)acetonitrile ≥96.0% (by GC)
(3-Iodophenyl)acetonitrile
Formula: C₈H₆IN
MW: 243.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00040890
CAS Number: 130723-54-5
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to lump
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR