4-Fluoro-3-toluoyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 4-Fluoro-3-methylbenzoyl chloride, 4-Fluoro-3-methylbenzene-1-carbonyl chloride

F1116-25G F1116-5G
TCF1116-25GEA 295 USD
TCF1116-25G TCF1116-5G
4-Fluoro-3-toluoyl chloride ≥98.0% (by GC, titration analysis)
4-Fluoro-3-toluoyl chloride
Formula: C₈H₆ClFO
Boiling Pt: 115 °C
Flash Pt: 98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631426
CAS Number: 455-84-5
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR