1-Fluoro-3-ethynylbenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: (3-Fluorophenyl)ethyne, 3-Fluorophenylacetylene, (3-Fluorophenyl)acetylene, 1-Ethynyl-3-fluorobenzene

E1332-25G E1332-5G
TCE1332-25GEA 359 USD
TCE1332-25G TCE1332-5G
1-Fluoro-3-ethynylbenzene ≥98.0% (by GC)
1-Fluoro-3-ethynylbenzene
Formula: C₈H₅F
Boiling Pt: 40 °C
Flash Pt: 32 °C
MDL Number: MFCD03093798
CAS Number: 2561-17-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR