3-Iodophenyl isothiocyanate ≥98.0% (by GC)

Supplier: TCI America
Danger

I0985-1G I0985-5G
TCI0985-1GEA 41 USD
TCI0985-1G TCI0985-5G
3-Iodophenyl isothiocyanate ≥98.0% (by GC)
3-Iodophenyl isothiocyanate
Formula: C₇H₄INS
MDL Number: MFCD00041074
CAS Number: 3125-73-3
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Melting point 45.0 to 49.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR