2-Fluoro-6-methoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

F1146-1G F1146-5G
TCF1146-1GEA 24 USD
TCF1146-1G TCF1146-5G
2-Fluoro-6-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
2-Fluoro-6-methoxybenzoic acid
Formula: C₈H₇FO₃
MDL Number: MFCD00671764
CAS Number: 137654-21-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 90.0 to 94.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR