2-Octyn-1-ol ≥98.0% (by GC)

Supplier: TCI America

O0493-25ML O0493-5ML
TCO0493-25MLEA 233 USD
TCO0493-25ML TCO0493-5ML
2-Octyn-1-ol ≥98.0% (by GC)
2-Octyn-1-ol
Formula: C₈H₁₄O
MW: 126.2 g/mol
Boiling Pt: 78 °C
Melting Pt: –18 °C
Flash Pt: 91 °C
MDL Number: MFCD00039542
CAS Number: 20739-58-6

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR