1,10-Phenanthroline ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

Synonyms: o-Phenanthroline

P2099-25G P2099-5G
TCP2099-25GEA 95 USD
TCP2099-25G TCP2099-5G
1,10-Phenanthroline ≥98.0% (by HPLC, titration analysis)
1,10-Phenanthroline
Formula: C₁₂H₈N₂
MDL Number: MFCD00011678
CAS Number: 66-71-7
EINECS: 200-629-2
UN: 2811
ADR: 6.1,III
Merck Index: 14,07214

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 118.0 to 122.0 deg-C
Water max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR