Tin(II) iodide ≥97.0% (by titrimetric analysis) for perovskite precursor
Supplier: TCI America
|
Danger
|
Synonyms:
Tin(II) iodide, Tin diiodide
T3449-1G
T3449-5G
TCT3449-1GEA
43
USD
TCT3449-1G
TCT3449-5G
Tin(II) iodide ≥97.0% (by titrimetric analysis) for perovskite precursor
Tin(II) iodide
Formula:
I₂Sn MW: 372.52 g/mol Boiling Pt: 714 Melting Pt: 320 |
MDL Number:
MFCD00049545 CAS Number: 10294-70-9 EINECS: 233-667-3 UN: 3260 ADR: 8 II Merck Index: 12,08942 |
Specification Test Results
Appearance | Light red to Yellow to Red powder to crystal |
Purity (Iodometric Titration) | min. 97.0 % |
Water (Vaporization methoed) | max. 100 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...