2-Isopropyl-5-methylanisole ≥96.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4-Isopropyl-3-methoxytoluene, 1-Isopropyl-2-methoxy-4-methylbenzene

I0996-25G I0996-5G
TCI0996-25GEA 144 USD
TCI0996-25G TCI0996-5G
2-Isopropyl-5-methylanisole ≥96.0% (by GC)
2-Isopropyl-5-methylanisole
Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Boiling Pt: 216 °C
Flash Pt: 51 °C
MDL Number: MFCD01674973
CAS Number: 1076-56-8
EINECS: 214-063-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR