4-Pentynoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

P2341-1G P2341-5G
TCP2341-1GEA 65 USD
TCP2341-1G TCP2341-5G
4-Pentynoic acid ≥98.0% (by GC, titration analysis)
4-Pentynoic acid
Formula: C₅H₆O₂
Boiling Pt: 204 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004407
CAS Number: 6089-09-4
EINECS: 228-028-0
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 55.0 to 59.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR