1-Ethynyl-3-nitrobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3-Nitrophenylacetylene, 1-Ethynyl-3-nitrobenzene, (3-Nitrophenyl)ethyne, (3-Nitrophenyl)acetylene

N1148-1G N1148-200MG
TCN1148-1GEA 107 USD
TCN1148-1G TCN1148-200MG
1-Ethynyl-3-nitrobenzene ≥98.0% (by GC)
1-Ethynyl-3-nitrobenzene
Formula: C₈H₅NO₂
MW: 147.13
Boiling Pt: 120 °C
MDL Number: MFCD02181150
CAS Number: 3034-94-4
EINECS: 221-232-0

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR