1,5-Diaminonaphthalene ≥99.5% (by GC), purified by sublimation
Supplier: TCI America
|
Warning
|
Synonyms:
1,5-naphthylenediamine, 1,5-Naphthalenediamine
D5689-1G
D5689-5G
TCD5689-1GEA
98
USD
TCD5689-1G
TCD5689-5G
1,5-Diaminonaphthalene ≥99.5% (by GC), purified by sublimation
1,5-Diaminonaphthalene
Formula:
C₁₀H₁₀N₂ MW: 158.2 g/mol |
MDL Number:
MFCD00004029 CAS Number: 2243-62-1 EINECS: 218-817-8 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 99.5 % |
Purity (Nonaqueous Titration) | min. 99.0 % |
Melting point | 189.0 to 194.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...