1,2-Bis(diethylamino)ethane ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: N1,N1,N2,N2-Tetraethylethane-1,2-diamine, N,N,N',N'-Tetraethylethylenediamine

T3371-5ML T3371-25ML
TCT3371-5MLEA 40.1 USD
TCT3371-5ML TCT3371-25ML
1,2-Bis(diethylamino)ethane ≥98.0% (by GC, titration analysis)
1,2-Bis(diethylamino)ethane
Formula: C₁₀H₂₄N₂
MW: 172.31 g/mol
Boiling Pt: 190 °C
Flash Pt: 58 °C
MDL Number: MFCD00009055
CAS Number: 150-77-6
EINECS: 205-770-3
UN: 2734
ADR: 8,II

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR