(3-Iodopropyl)trimethoxysilane ≥98.0% (by GC)

Supplier: TCI America
Warning

I1106-25G I1106-5G
TCI1106-25GEA 525 USD
TCI1106-25G TCI1106-5G
(3-Iodopropyl)trimethoxysilane ≥98.0% (by GC)
(3-Iodopropyl)trimethoxysilane
Formula: IH₂CCH₂CH₂Si(OCH₃)₃
MW: 290.17 g/mol
Boiling Pt: 80 °C
Flash Pt: 78 °C
MDL Number: MFCD00054838
CAS Number: 14867-28-8

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR