4-Fluorophthalonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

F1169-1G F1169-5G
TCF1169-1GEA 133 USD
TCF1169-1G TCF1169-5G
4-Fluorophthalonitrile ≥98.0% (by GC)
4-Fluorophthalonitrile
Formula: C₈H₃FN₂
MDL Number: MFCD00210430
CAS Number: 65610-14-2
UN: 3439
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 101.0 to 105.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR