Phenyl anthranilate ≥98.0% (by GC)

Supplier: TCI America
Warning

P2400-1G P2400-5G
TCP2400-1GEA 96 USD
TCP2400-1G TCP2400-5G
Phenyl anthranilate ≥98.0% (by GC)
Phenyl anthranilate
Formula: C₁₃H₁₁NO₂
MDL Number: MFCD00060603
CAS Number: 10268-69-6
EINECS: 233-608-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 70.0 to 74.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR