4-Vinylpyridine ≥95.0% (by GC) stabilized

Supplier: TCI America
Danger


Stabilization: Stabilised with hydroquinone (HQ)

V0150-25ML V0150-500ML
TCV0150-25MLEA 41 USD
TCV0150-25ML TCV0150-500ML
4-Vinylpyridine ≥95.0% (by GC) stabilized
4-Vinylpyridine
Formula: C₇H₇N
MW: 105.14 g/mol
Boiling Pt: 59 °C
Flash Pt: 56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006447
CAS Number: 100-43-6
EINECS: 202-852-0
UN: 3073
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Colourless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR